EMAIL THIS PAGE TO A FRIEND
PS2017 Supelco

Omethoate

analytical standard

  • CAS Number 1113-02-6

  • Empirical Formula (Hill Notation) C5H12NO4PS

  • Molecular Weight 213.19

  •  Beilstein/REAXYS Number 1785256

  •  EC Number 214-197-8

  •  MDL number MFCD00055441

Purchase

Properties

grade   analytical standard
InChI Key   PZXOQEXFMJCDPG-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2017
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1/PG 2
WGK Germany 
3
RTECS 
TF8050000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?