EMAIL THIS PAGE TO A FRIEND
PS2024 Supelco

Sulfotep

analytical standard

Synonym: O,O,O′,O′-Tetraethyl dithiopyrophosphate

  • CAS Number 3689-24-5

  • Empirical Formula (Hill Notation) C8H20O5P2S2

  • Molecular Weight 322.32

  •  Beilstein Registry Number 1714019

  •  EC Number 222-995-2

  •  MDL number MFCD00026764

  •  PubChem Substance ID 24899135

Purchase

Properties

grade   analytical standard
InChI Key   XIUROWKZWPIAIB-UHFFFAOYSA-N
form   neat
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., PS-2024
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2810 - class 6.1 - PG 1 - MP - RQ - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
XN4375000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?