EMAIL THIS PAGE TO A FRIEND
PS2034 Supelco

Cyanophos

analytical standard

Synonym: O-(4-Cyanophenyl) O,O-dimethyl phosphorothioate

  • CAS Number 2636-26-2

  • Empirical Formula (Hill Notation) C9H10NO3PS

  • Molecular Weight 243.22

  •  Beilstein Registry Number 2695901

  •  EC Number 220-130-3

  •  MDL number MFCD00055485

  •  PubChem Substance ID 24899139

Purchase

Properties

grade   analytical standard
InChI Key   SCKHCCSZFPSHGR-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2034
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3278 6.1 / PGIII
WGK Germany 
3
RTECS 
TF7600000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?