EMAIL THIS PAGE TO A FRIEND
PS2040

Acetochlor

analytical standard

  • CAS Number 34256-82-1

  • Empirical Formula (Hill Notation) C14H20ClNO2

  • Molecular Weight 269.77

  •  Beilstein/REAXYS Number 2859702

  •  EC Number 251-899-3

  •  MDL number MFCD00143534

  •  PubChem Substance ID 24899141

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2040
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   CCOCN(C(=O)CCl)c1c(C)cccc1CC
InChI   1S/C14H20ClNO2/c1-4-12-8-6-7-11(3)14(12)16(10-18-5-2)13(17)9-15/h6-8H,4-5,9-10H2,1-3H3
InChI key   VTNQPKFIQCLBDU-UHFFFAOYSA-N

Description

General description

Acetochlor belongs to the class of chloroacetanilide herbicides. It is primarily used as a pre-emergence herbicide for a wide range of crops.

Application

Acetochlor may be used as a reference standard for the determination of acetochlor in water samples using high-performance liquid chromatography-diode array detection (HPLC-DAD) and high-performance liquid chromatography/mass spectrometry (HPLC/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
RTECS 
AB5457000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?