EMAIL THIS PAGE TO A FRIEND
PS2047 Supelco

Dichlofluanid

analytical standard

  • CAS Number 1085-98-9

  • Empirical Formula (Hill Notation) C9H11Cl2FN2O2S2

  • Molecular Weight 333.23

  •  Beilstein Registry Number 2947992

  •  EC Number 214-118-7

  •  MDL number MFCD00078639

  •  PubChem Substance ID 24899146

Purchase

Properties

grade   analytical standard
InChI Key   WURGXGVFSMYFCG-UHFFFAOYSA-N
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-2047
Featured Industry   Agriculture
Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
WO6475000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?