EMAIL THIS PAGE TO A FRIEND
PS2072 Supelco

Fenbuconazol

analytical standard

  • CAS Number 114369-43-6

  • Empirical Formula (Hill Notation) C19H17ClN4

  • Molecular Weight 336.82

  •  Beilstein/REAXYS Number 8333667

  •  EC Number 406-140-2

  •  MDL number MFCD01632336

  •  PubChem Substance ID 24899160

Purchase

Properties

grade   analytical standard
InChI Key   RQDJADAKIFFEKQ-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2072
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
XZ5257500

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?