EMAIL THIS PAGE TO A FRIEND
PS2079 Supelco

Benzoylprop-ethyl

analytical standard

  • CAS Number 22212-55-1

  • Empirical Formula (Hill Notation) C18H17Cl2NO3

  • Molecular Weight 366.24

  •  Beilstein Registry Number 2899940

  •  EC Number 244-845-5

  •  MDL number MFCD00143676

  •  PubChem Substance ID 24899163

Purchase

Properties

grade   analytical standard
InChI Key   SLCGUGMPSUYJAY-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2079
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
UE7550000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?