• USA Home
  • PS215 - Phenylmercuric hydroxide

EMAIL THIS PAGE TO A FRIEND
PS215 Supelco

Phenylmercuric hydroxide

analytical standard

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-215
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat
SMILES string   O[Hg]c1ccccc1
InChI   1S/C6H5.Hg.H2O/c1-2-4-6-5-3-1;;/h1-5H;;1H2/q;+1;/p-1
InChI key   NWDUDBMLUKJHGK-UHFFFAOYSA-M

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 1894 6.1 / PGII
WGK Germany 
3
RTECS 
OW4940000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?