EMAIL THIS PAGE TO A FRIEND
PS22

Maneb

analytical standard

Synonym: Manganese ethylenebis(dithiocarbamate)

Purchase

Properties

grade   analytical standard
form   neat
mol wt   Mw 265.30 (as monomer)
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-22
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   S=C1NCCNC(=S)S[Mn]S1
InChI   1S/C4H8N2S4.Mn/c7-3(8)5-1-2-6-4(9)10;/h1-2H2,(H2,5,7,8)(H2,6,9,10);/q;+2/p-2
InChI key   YKSNLCVSTHTHJA-UHFFFAOYSA-L

Description

General description

Maneb is a fungicide, which finds applications in agriculture to control the early and late blights in potatoes and tomatoes.

Application

Maneb may be used as a reference standard in the determination of maneb in tomato samples using liquid chromatography coupled with ultraviolet and diode array detector (LC-UV-DAD)

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2210 4.3(4.2) / PGIII
WGK Germany 
1
RTECS 
OP0700000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?