EMAIL THIS PAGE TO A FRIEND
PS230

Thiophanate-methyl

analytical standard

  • CAS Number 23564-05-8

  • Empirical Formula (Hill Notation) C12H14N4O4S2

  • Molecular Weight 342.39

  •  Beilstein/REAXYS Number 937942

  •  EC Number 245-740-7

  •  MDL number MFCD00055294

  •  PubChem Substance ID 24899171

Purchase

Properties

grade   analytical standard
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-230
Featured Industry   Agriculture
Cleaning Products
Cosmetics
Environmental
Food and Beverages
Personal Care
SMILES string   COC(=O)NC(=S)Nc1ccccc1NC(=S)NC(=O)OC
InChI   1S/C12H14N4O4S2/c1-19-11(17)15-9(21)13-7-5-3-4-6-8(7)14-10(22)16-12(18)20-2/h3-6H,1-2H3,(H2,13,15,17,21)(H2,14,16,18,22)
InChI key   QGHREAKMXXNCOA-UHFFFAOYSA-N

Description

General description

Thiophanate-methyl is a fungicide, which is commonly used in the cultivation of fruits and vegetables.

Application

Thiophanate-methyl standard may be used in the determination of thiophanate-methyl in vegetable samples using microwave assisted extraction (MAE), coupled with high performance liquid chromatography (HPLC). It may also be used as a reference standard in the analysis of thiophanate-methyl in fruit samples using high performance liquid chromatography (HPLC) coupled with diode array detector (DAD).

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
BA3675000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?