• USA Home
  • PS24 - Tetramethylthiuram disulfide

EMAIL THIS PAGE TO A FRIEND
PS24 Supelco

Tetramethylthiuram disulfide

analytical standard

Synonym: Bis(dimethylthiocarbamoyl) disulfide, Bis(dimethylthiocarbamyl) disulfide, Thiram

  • CAS Number 137-26-8

  • Linear Formula (CH3)2NCSS2CSN(CH3)2

  • Molecular Weight 240.43

  •  Beilstein/REAXYS Number 1725821

  •  EC Number 205-286-2

  •  MDL number MFCD00008325

  •  PubChem Substance ID 24899172

Purchase

Properties

grade   analytical standard
form   neat
autoignition temp.   316 °F
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-24
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   156-158 °C (lit.)
Featured Industry   Agriculture
Cleaning Products
Cosmetics
Environmental
Food and Beverages
Personal Care
format   neat
SMILES string   CN(C)C(=S)SSC(=S)N(C)C
InChI   1S/C6H12N2S4/c1-7(2)5(9)11-12-6(10)8(3)4/h1-4H3
InChI key   KUAZQDVKQLNFPE-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
JO1400000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?