EMAIL THIS PAGE TO A FRIEND
PS307 Supelco

MCPB

analytical standard

Synonym: 4-(4-Chloro-o-tolyloxy)butyric acid, 4-(4-Chloro-2-methylphenoxy)butyric acid

  • CAS Number 94-81-5

  • Empirical Formula (Hill Notation) C11H13ClO3

  • Molecular Weight 228.67

  •  Beilstein/REAXYS Number 2215202

  •  EC Number 202-365-3

  •  MDL number MFCD00002820

  •  PubChem Substance ID 24899181

Purchase

Properties

Related Categories All Herbicide Standards, Analytical Standards, Analytical/Chromatography, E-M, Herbicides,
grade   analytical standard
InChI Key   LLWADFLAOKUBDR-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-307
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
ES8575000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 6) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 6) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?