• USA Home
  • PS314 - 3-Amino-2,5-dichlorobenzoic acid

EMAIL THIS PAGE TO A FRIEND
PS314

3-Amino-2,5-dichlorobenzoic acid

analytical standard

Synonym: Chloramben

  • CAS Number 133-90-4

  • Linear Formula H2NC6H2(Cl)2CO2H

  • Molecular Weight 206.03

  •  Beilstein/REAXYS Number 2365906

  •  EC Number 205-123-5

  •  MDL number MFCD00065093

  •  PubChem Substance ID 24899182

Purchase

Properties

grade   analytical standard
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-314
mp   194-197 °C (dec.) (lit.)
Featured Industry   Agriculture
Environmental
SMILES string   Nc1cc(Cl)cc(C(O)=O)c1Cl
InChI   1S/C7H5Cl2NO2/c8-3-1-4(7(11)12)6(9)5(10)2-3/h1-2H,10H2,(H,11,12)
InChI key   HSSBORCLYSCBJR-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
WGK 3
RTECS 
DG1925000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?