EMAIL THIS PAGE TO A FRIEND
PS324 Supelco

Mecoprop

analytical standard

Synonym: 2-(4-Chloro-2-methylphenoxy)propionic acid

  • CAS Number 93-65-2

  • Empirical Formula (Hill Notation) C10H11ClO3

  • Molecular Weight 214.65

  •  Beilstein/REAXYS Number 6568283

  •  EC Number 202-264-4

  •  MDL number MFCD00002648

  •  PubChem Substance ID 24899183

Purchase

Properties

grade   analytical standard
InChI Key   WNTGYJSOUMFZEP-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-324
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat

Description

General description

Mecoprop is a chlorophenoxypropionic herbicide, used for the post-emergence control of cleavers, chickweed and other weeds in cereal crops.

Application

Mecoprop may be used as a reference standard for the determination of mecoprop in natural water by high-performance liquid chromatography (HPLC).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
2
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?