• USA Home
  • PS343 - N-1-Naphthylphthalamidic acid

EMAIL THIS PAGE TO A FRIEND
PS343 Supelco

N-1-Naphthylphthalamidic acid

analytical standard

Synonym: N-(1-Naphthyl)phthalamidic acid, Naptalam

  • CAS Number 132-66-1

  • Empirical Formula (Hill Notation) C18H13NO3

  • Molecular Weight 291.30

  •  Beilstein/REAXYS Number 2814102

  •  MDL number MFCD00037725

  •  PubChem Substance ID 24899188

Purchase

Properties

grade   analytical standard
InChI Key   JXTHEWSKYLZVJC-UHFFFAOYSA-N
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-343
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

General description

N-1-Naphthylphthalamidic acid is a polar auxin transport inhibitor.

Application

N-1-Naphthylphthalamidic acid may be used as a model compound to demonstrate the effect of N-1-naphthylphthalamidic acid in maize. It disrupts leaf initiation, KNOX protein regulation, and formation of leaf margins in maize.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?