EMAIL THIS PAGE TO A FRIEND
PS345 Supelco

Endothal monohydrate

analytical standard

Synonym: 3,6-Epoxy-cyclohexan-1,2-dicarboxylic acid monohydrate

  • CAS Number 62059-43-2

  • Empirical Formula (Hill Notation) C8H10O5 · H2O

  • Molecular Weight 204.18

  •  MDL number MFCD01631318

  •  PubChem Substance ID 24899189

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc. PS-345
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat
SMILES string   O.OC(=O)C1[C@H]2CC[C@H](O2)C1C(O)=O
InChI   1S/C8H10O5.H2O/c9-7(10)5-3-1-2-4(13-3)6(5)8(11)12;/h3-6H,1-2H2,(H,9,10)(H,11,12);1H2/t3-,4+,5?,6?;
InChI key   RHLALKDQINFLPM-UCDWXUIISA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?