EMAIL THIS PAGE TO A FRIEND
PS357

Alachlor

analytical standard

Synonym: 2-Chloro-2′,6′-diethyl-N-(methoxymethyl)acetanilide

  • CAS Number 15972-60-8

  • Empirical Formula (Hill Notation) C14H20ClNO2

  • Molecular Weight 269.77

  •  Beilstein/REAXYS Number 2944476

  •  EC Number 240-110-8

  •  MDL number MFCD00041817

  •  PubChem Substance ID 24899194

Purchase

Properties

grade   analytical standard
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-357
Featured Industry   Agriculture
Environmental
SMILES string   CCc1cccc(CC)c1N(COC)C(=O)CCl
InChI   1S/C14H20ClNO2/c1-4-11-7-6-8-12(5-2)14(11)16(10-18-3)13(17)9-15/h6-8H,4-5,9-10H2,1-3H3
InChI key   XCSGPAVHZFQHGE-UHFFFAOYSA-N

Description

General description

Alachlor is a carcinogenic herbicide.

Application

Alachlor may be used as a reference standard for the determination of alachlor in water by solid-phase extraction and enzyme-linked immunosorbent assay.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
AE1225000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?