EMAIL THIS PAGE TO A FRIEND
PS381 Supelco

Simetryn

analytical standard

Synonym: 2,4-Bis(ethylamino)-6-methylthio-1,3,5-triazine

  • CAS Number 1014-70-6

  • Empirical Formula (Hill Notation) C8H15N5S

  • Molecular Weight 213.30

  •  Beilstein/REAXYS Number 11728

  •  EC Number 213-801-7

  •  MDL number MFCD00078720

  •  PubChem Substance ID 24899207

Purchase

Properties

grade   analytical standard
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., PS-381
Featured Industry   Agriculture
SMILES string   CCNc1nc(NCC)nc(SC)n1
InChI   1S/C8H15N5S/c1-4-9-6-11-7(10-5-2)13-8(12-6)14-3/h4-5H2,1-3H3,(H2,9,10,11,12,13)
InChI key   MGLWZSOBALDPEK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
XY4025000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?