• USA Home
  • PS392 - 2,6-Dichlorobenzonitrile

EMAIL THIS PAGE TO A FRIEND
PS392 Supelco

2,6-Dichlorobenzonitrile

analytical standard

Synonym: Dichlobenil

Purchase

Properties

grade   analytical standard
InChI Key   YOYAIZYFCNQIRF-UHFFFAOYSA-N
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-392
mp   143-146 °C (lit.)
Featured Industry   Agriculture
Environmental
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
DI3500000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?