EMAIL THIS PAGE TO A FRIEND
PS395

Atraton

analytical standard

Synonym: Atrazine-methoxy

  • CAS Number 1610-17-9

  • Empirical Formula (Hill Notation) C9H17N5O

  • Molecular Weight 211.26

  •  Beilstein/REAXYS Number 613098

  •  EC Number 216-547-5

  •  MDL number MFCD00078647

  •  PubChem Substance ID 24899218

Purchase

Properties

grade   analytical standard
packaging   ampule of 500 mg
mfr. no.   Chem Service, Inc., PS-395
Featured Industry   Agriculture
Environmental
SMILES string   CCNc1nc(NC(C)C)nc(OC)n1
InChI   1S/C9H17N5O/c1-5-10-7-12-8(11-6(2)3)14-9(13-7)15-4/h6H,5H2,1-4H3,(H2,10,11,12,13,14)
InChI key   PXWUKZGIHQRDHL-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
XY8750000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?