EMAIL THIS PAGE TO A FRIEND
PS398 Supelco

Metribuzin

analytical standard

  • CAS Number 21087-64-9

  • Empirical Formula (Hill Notation) C8H14N4OS

  • Molecular Weight 214.29

  •  Beilstein Registry Number 746650

  •  EC Number 244-209-7

  •  MDL number MFCD00055525

Purchase

Properties

grade   analytical standard
InChI Key   FOXFZRUHNHCZPX-UHFFFAOYSA-N
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-398
Featured Industry   Agriculture
Environmental
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
XZ2990000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?