EMAIL THIS PAGE TO A FRIEND
PS406 Supelco

Isopropalin

analytical standard

Synonym: 4-Isopropyl-2,6-dinitro-N,N-dipropylaniline

  • CAS Number 33820-53-0

  • Empirical Formula (Hill Notation) C15H23N3O4

  • Molecular Weight 309.36

  •  Beilstein Registry Number 2762577

  •  EC Number 251-690-7

  •  MDL number MFCD00055306

  •  PubChem Substance ID 329823656

Purchase

Properties

Related Categories All Herbicide Standards, Analytical Standards, Analytical/Chromatography, Dinitroaniline, E-M,
grade   analytical standard
InChI Key   NEKOXWSIMFDGMA-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-406
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
BX9286000
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?