EMAIL THIS PAGE TO A FRIEND
PS407

Tebuthiuron

analytical standard

Synonym: TBH

  • CAS Number 34014-18-1

  • Empirical Formula (Hill Notation) C9H16N4OS

  • Molecular Weight 228.31

  •  Beilstein/REAXYS Number 527479

  •  EC Number 251-793-7

  •  MDL number MFCD00078732

  •  PubChem Substance ID 24899221

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-407
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   CNC(=O)N(C)c1nnc(s1)C(C)(C)C
InChI   1S/C9H16N4OS/c1-9(2,3)6-11-12-8(15-6)13(5)7(14)10-4/h1-5H3,(H,10,14)
InChI key   HBPDKDSFLXWOAE-UHFFFAOYSA-N

Description

General description

Tebuthiuron is a herbicide used for weed control management.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Tebuthiuron may be used as a reference standard for the analysis of tebuthiuron herbicide in soil samples and plant samples by high-performance liquid chromatography (HPLC) method. and gas chromatographic technique coupled with flame photometric detector (GC-FPD), respectively.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
YS4250000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?