EMAIL THIS PAGE TO A FRIEND
PS44

Dichlorprop

analytical standard

Synonym: 2-(2,4-Dichlorophenoxy)propionic acid

  • CAS Number 120-36-5

  • Empirical Formula (Hill Notation) C9H8Cl2O3

  • Molecular Weight 235.06

  •  Beilstein/REAXYS Number 6568822

  •  EC Number 204-390-5

  •  MDL number MFCD00002645

  •  PubChem Substance ID 24899232

Purchase

Properties

grade   analytical standard
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-44
Featured Industry   Agriculture
Environmental
SMILES string   CC(Oc1ccc(Cl)cc1Cl)C(O)=O
InChI   1S/C9H8Cl2O3/c1-5(9(12)13)14-8-3-2-6(10)4-7(8)11/h2-5H,1H3,(H,12,13)
InChI key   MZHCENGPTKEIGP-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
UF1050000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?