EMAIL THIS PAGE TO A FRIEND
PS501

Molinate

analytical standard

  • CAS Number 2212-67-1

  • Empirical Formula (Hill Notation) C9H17NOS

  • Molecular Weight 187.30

  •  Beilstein/REAXYS Number 1239196

  •  EC Number 218-661-0

  •  MDL number MFCD00055352

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-501
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Cleaning Products
Cosmetics
Environmental
Food and Beverages
Personal Care
format   neat
SMILES string   CCSC(=O)N1CCCCCC1
InChI   1S/C9H17NOS/c1-2-12-9(11)10-7-5-3-4-6-8-10/h2-8H2,1H3
InChI key   DEDOPGXGGQYYMW-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
CM2625000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?