EMAIL THIS PAGE TO A FRIEND
PS506 Supelco

Tri-allate

analytical standard

Synonym: S-(2,3,3-Trichloroallyl) N,N-diisopropylthiocarbamate, Triallat

  • CAS Number 2303-17-5

  • Empirical Formula (Hill Notation) C10H16Cl3NOS

  • Molecular Weight 304.66

  •  Beilstein Registry Number 1875853

  •  EC Number 218-962-7

  •  MDL number MFCD00078730

  •  PubChem Substance ID 24899236

Purchase

Properties

grade   analytical standard
InChI Key   MWBPRDONLNQCFV-UHFFFAOYSA-N
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-506
Featured Industry   Agriculture

Description

Application

Trillate may be used as reference standard in the determination of trillate in hemodialysis saline solutions using high performance liquid chromatography coupled with ultraviolet detector (HPLC-UV).

General description

Triallate is a selective preemergence and postemergence type of herbicide, which is commonly applied to soil to control the growth of annual grasses in crop cultivation. Its mode of action involves the inhibition of cell division, cell elongation, lipid biosynthesis and fatty acids.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
EZ8575000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?