EMAIL THIS PAGE TO A FRIEND
PS543 Supelco

Methiocarb

analytical standard

Synonym: Mercaptodimethur

  • CAS Number 2032-65-7

  • Empirical Formula (Hill Notation) C11H15NO2S

  • Molecular Weight 225.31

  •  Beilstein/REAXYS Number 1881431

  •  EC Number 217-991-2

  •  MDL number MFCD00055450

Purchase

Properties

grade   analytical standard
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-543
Featured Industry   Agriculture
Environmental
SMILES string   CNC(=O)Oc1cc(C)c(SC)c(C)c1
InChI   1S/C11H15NO2S/c1-7-5-9(14-11(13)12-3)6-8(2)10(7)15-4/h5-6H,1-4H3,(H,12,13)
InChI key   YFBPRJGDJKVWAH-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 2 - MP - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
FC5775000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?