• USA Home
  • PS601 - Tetraethyl pyrophosphate

EMAIL THIS PAGE TO A FRIEND
PS601

Tetraethyl pyrophosphate

analytical standard

Synonym: Tetraethyl diphosphate

  • CAS Number 107-49-3

  • Empirical Formula (Hill Notation) C8H20O7P2

  • Molecular Weight 290.19

  •  Beilstein/REAXYS Number 1714017

  •  EC Number 203-495-3

  •  MDL number MFCD00055459

  •  PubChem Substance ID 24899248

Purchase

Properties

grade   analytical standard
packaging   ampule of 500 mg
mfr. no.   Chem Service, Inc., PS-601
Featured Industry   Agriculture
Cleaning Products
Cosmetics
Environmental
Food and Beverages
Personal Care
SMILES string   CCOP(=O)(OCC)OP(=O)(OCC)OCC
InChI   1S/C8H20O7P2/c1-5-11-16(9,12-6-2)15-17(10,13-7-3)14-8-4/h5-8H2,1-4H3
InChI key   IDCBOTIENDVCBQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2810 - class 6.1 - PG 1 - MP - RQ - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
UX6825000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?