EMAIL THIS PAGE TO A FRIEND
PS602

Dicrotophos

analytical standard

  • CAS Number 141-66-2

  • Empirical Formula (Hill Notation) C8H16NO5P

  • Molecular Weight 237.19

  •  Beilstein/REAXYS Number 1880084

  •  EC Number 205-494-3

  •  MDL number MFCD00072499

  •  PubChem Substance ID 24899249

Purchase

Properties

grade   analytical standard
form   neat
packaging   pkg of 250 mg
mfr. no.   Chem Service, Inc., PS-602
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   COP(=O)(OC)O\C(C)=C\C(=O)N(C)C
InChI   1S/C8H16NO5P/c1-7(6-8(10)9(2)3)14-15(11,12-4)13-5/h6H,1-5H3/b7-6+
InChI key   VEENJGZXVHKXNB-VOTSOKGWSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
3
RTECS 
TC3850000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?