EMAIL THIS PAGE TO A FRIEND
PS612 Supelco

Fenamiphos

analytical standard

Synonym: Phenamiphos

  • CAS Number 22224-92-6

  • Empirical Formula (Hill Notation) C13H22NO3PS

  • Molecular Weight 303.36

  •  Beilstein Registry Number 4752893

  •  EC Number 244-848-1

  •  MDL number MFCD00055454

  •  PubChem Substance ID 24899256

Purchase

Properties

grade   analytical standard
InChI Key   ZCJPOPBZHLUFHF-UHFFFAOYSA-N
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-612
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
TB3675000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?