• USA Home
  • PS613 - Dimethyl p-nitrophenylphosphate

EMAIL THIS PAGE TO A FRIEND
PS613 Supelco

Dimethyl p-nitrophenylphosphate

analytical standard

Synonym: O,O-Dimethyl O-(4-nitrophenyl) phosphate, Paraoxon-methyl

  • CAS Number 950-35-6

  • Empirical Formula (Hill Notation) C8H10NO6P

  • Molecular Weight 247.14

  •  Beilstein/REAXYS Number 1884904

  •  MDL number MFCD00055531

  •  PubChem Substance ID 24899257

Purchase

Properties

grade   analytical standard
InChI Key   BAFQDKPJKOLXFZ-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-613
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Petroleum
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGI
WGK Germany 
3
RTECS 
TC5250000
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?