EMAIL THIS PAGE TO A FRIEND
PS657

Fenchlorazol-ethyl

analytical standard

Synonym: Ethyl 1-(2,4-dichlorophenyl)-5-(trichloromethyl)-1H-1,2,4-triazole-3-carboxylate

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-657
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Petroleum
format   neat
SMILES string   CCOC(=O)c1nc(n(n1)-c2ccc(Cl)cc2Cl)C(Cl)(Cl)Cl
InChI   1S/C12H8Cl5N3O2/c1-2-22-10(21)9-18-11(12(15,16)17)20(19-9)8-4-3-6(13)5-7(8)14/h3-5H,2H2,1H3
InChI key   GMBRUAIJEFRHFQ-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
XZ4272500

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?