EMAIL THIS PAGE TO A FRIEND
PS667 Supelco

Fensulfothion

analytical standard

Synonym: Diethyl 4-(methylthio)phenyl phosphate

  • CAS Number 115-90-2

  • Empirical Formula (Hill Notation) C11H17O4PS2

  • Molecular Weight 308.35

  •  Beilstein/REAXYS Number 2219515

  •  EC Number 204-114-3

  •  MDL number MFCD00055487

  •  PubChem Substance ID 24899267

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Insecticides, Nematicides, Organophorous,
grade   analytical standard
InChI Key   XDNBJTQLKCIJBV-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-667
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGI
WGK Germany 
3
RTECS 
TF3850000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 1) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 1) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

GC Analysis of Organophosphorus Pesticides (OP-Pest) in Cabbage, Onion, and Mushrooms on SLB-5ms after SPE using Supelclean™ ENVI-Carb™ II/PSA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Organophosphorus Pesticides (OP-Pest) in Cabbage, Onion, and Mushrooms on SLB-5ms after SPE using Supelclean™ ENVI-Carb™ II/PSA
Keywords: Agriculture, Centrifugation, Chromatography, Degradations, Environmental, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification, Sample preparations, Separation, Solid phase extractions

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, MS Detection

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, MS Detection
Keywords: Chromatography, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, NPD Detection

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, NPD Detection
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-608
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?