EMAIL THIS PAGE TO A FRIEND
PS677

Leptophos

analytical standard

Synonym: O-(4-Bromo-2,5-dichlorophenyl) O-methyl phenylphosphonothioate

  • CAS Number 21609-90-5

  • Empirical Formula (Hill Notation) C13H10BrCl2O2PS

  • Molecular Weight 412.07

  •  Beilstein/REAXYS Number 2152663

  •  EC Number 244-472-8

  •  MDL number MFCD00128037

  •  PubChem Substance ID 24899271

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Insecticides, Organophorous, Pesticides & Metabolites Standards More...
grade   analytical standard
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-677
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat
SMILES string   COP(=S)(Oc1cc(Cl)c(Br)cc1Cl)c2ccccc2
InChI   1S/C13H10BrCl2O2PS/c1-17-19(20,9-5-3-2-4-6-9)18-13-8-11(15)10(14)7-12(13)16/h2-8H,1H3
InChI key   CVRALZAYCYJELZ-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN3464 - class 6.1 - PG 2 - EHS - Organophosphorus compound, toxic, solid, n.o.s., HI: all (not BR)
WGK Germany 
WGK 3
RTECS 
TB1720000
Flash Point(F) 
212.0 °F
Flash Point(C) 
> 100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 6) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 6) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, MS Detection

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, MS Detection
Keywords: Chromatography, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, NPD Detection

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, NPD Detection
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on SPB®-608
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

replacement part

Product #

Description

Add to Cart

33366 Leptophos, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?