EMAIL THIS PAGE TO A FRIEND
PS680

Terbufos

analytical standard

  • CAS Number 13071-79-9

  • Empirical Formula (Hill Notation) C9H21O2PS3

  • Molecular Weight 288.43

  •  Beilstein/REAXYS Number 1710115

  •  EC Number 235-963-8

  •  MDL number MFCD00055355

  •  PubChem Substance ID 24899273

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 500 mg
mfr. no.   Chem Service, Inc., PS-680
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   CCOP(=S)(OCC)SCSC(C)(C)C
InChI   1S/C9H21O2PS3/c1-6-10-12(13,11-7-2)15-8-14-9(3,4)5/h6-8H2,1-5H3
InChI key   XLNZEKHULJKQBA-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2810 6.1 / PGI
WGK Germany 
WGK 3
RTECS 
TD7200000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?