EMAIL THIS PAGE TO A FRIEND
PS681

Isazophos

analytical standard

  • CAS Number 42509-80-8

  • Empirical Formula (Hill Notation) C9H17ClN3O3PS

  • Molecular Weight 313.74

  •  Beilstein/REAXYS Number 8145294

  •  EC Number 255-863-8

  •  MDL number MFCD00078699

  •  PubChem Substance ID 24899274

Purchase

Properties

grade   analytical standard
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-681
Featured Industry   Agriculture
Environmental
SMILES string   CCOP(=S)(OCC)Oc1nc(Cl)n(n1)C(C)C
InChI   1S/C9H17ClN3O3PS/c1-5-14-17(18,15-6-2)16-9-11-8(10)13(12-9)7(3)4/h7H,5-6H2,1-4H3
InChI key   XRHGWAGWAHHFLF-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3278 - class 6.1 - PG 2 - EHS - Organophosphorus compound, toxic, liquid, n.o.s., HI: all (not BR)
WGK Germany 
3
RTECS 
TE7760000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?