EMAIL THIS PAGE TO A FRIEND
PS701 Supelco

Kepone

analytical standard

Synonym: Chlordecone, Perchloropentacyclo[5.3.0.02,6.03,9.04,8]decan-5-one

Purchase

Properties

grade   analytical standard
InChI Key   LHHGDZSESBACKH-UHFFFAOYSA-N
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc., PS-701
Featured Industry   Agriculture
Cosmetics, Personal Care, and Cleaning Products
Environmental
Food and Beverages
Gene Information   rat ... Ar(24208)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
PC8575000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?