EMAIL THIS PAGE TO A FRIEND
PS738 Supelco

Acephate

analytical standard

Synonym: O,S-Dimethyl N-acetylphosphoramidothioate

  • CAS Number 30560-19-1

  • Empirical Formula (Hill Notation) C4H10NO3PS

  • Molecular Weight 183.17

  •  Beilstein/REAXYS Number 1936365

  •  EC Number 250-241-2

  •  MDL number MFCD00055361

  •  PubChem Substance ID 24899286

Purchase

Properties

grade   analytical standard
form   neat
packaging   pkg of 250 mg
mfr. no.   Chem Service, Inc., PS-738
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   COP(=O)(NC(C)=O)SC
InChI   1S/C4H10NO3PS/c1-4(6)5-9(7,8-2)10-3/h1-3H3,(H,5,6,7)
InChI key   YASYVMFAVPKPKE-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
2
RTECS 
TB4760000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?