EMAIL THIS PAGE TO A FRIEND
PS739

Bendiocarb

analytical standard

  • CAS Number 22781-23-3

  • Empirical Formula (Hill Notation) C11H13NO4

  • Molecular Weight 223.23

  •  Beilstein/REAXYS Number 1315404

  •  EC Number 245-216-8

  •  MDL number MFCD00078629

  •  PubChem Substance ID 24899287

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-739
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat
SMILES string   CNC(=O)Oc1cccc2OC(C)(C)Oc12
InChI   1S/C11H13NO4/c1-11(2)15-8-6-4-5-7(9(8)16-11)14-10(13)12-3/h4-6H,1-3H3,(H,12,13)
InChI key   XEGGRYVFLWGFHI-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
FC1140000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?