EMAIL THIS PAGE TO A FRIEND
PS73 Supelco

Perthan

analytical standard

  • CAS Number 72-56-0

  • Empirical Formula (Hill Notation) C18H20Cl2

  • Molecular Weight 307.26

  •  Beilstein/REAXYS Number 2054366

  •  EC Number 200-785-1

  •  MDL number MFCD00045254

  •  PubChem Substance ID 24899282

Purchase

Properties

grade   analytical standard
InChI Key   QFMDFTQOJHFVNR-UHFFFAOYSA-N
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-73
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
KH5790000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

45615 Perthan, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?