EMAIL THIS PAGE TO A FRIEND
PS751A Supelco

Aminocarb

analytical standard

Synonym: (4-Dimethylamino-3-methylphenyl) N-methylcarbamate

  • CAS Number 2032-59-9

  • Empirical Formula (Hill Notation) C11H16N2O2

  • Molecular Weight 208.26

  •  Beilstein/REAXYS Number 2808681

  •  EC Number 217-990-7

  •  MDL number MFCD00055440

  •  PubChem Substance ID 24899288

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-751
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat
SMILES string   CNC(=O)Oc1ccc(N(C)C)c(C)c1
InChI   1S/C11H16N2O2/c1-8-7-9(15-11(14)12-2)5-6-10(8)13(3)4/h5-7H,1-4H3,(H,12,14)
InChI key   IMIDOCRTMDIQIJ-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
FC0175000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?