EMAIL THIS PAGE TO A FRIEND
PS757 Supelco

Pirimicarb

analytical standard

Synonym: 2-Dimethylamino-5,6-dimethyl-4-pyrimidinyl dimethylcarbamate

  • CAS Number 23103-98-2

  • Empirical Formula (Hill Notation) C11H18N4O2

  • Molecular Weight 238.29

  •  Beilstein Registry Number 663442

  •  EC Number 245-430-1

  •  MDL number MFCD00055520

  •  PubChem Substance ID 24899289

Purchase

Properties

grade   analytical standard
InChI Key   YFGYUFNIOHWBOB-UHFFFAOYSA-N
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-757
Featured Industry   Agriculture

Description

Application

Pirimicarb may be used as a reference standard for the determination of N-methylcarbamate pesticides in food by accelerated solvent extraction (ASE) and HPLC with post-column fluorescence.

General description

Pirimicarb is an insecticide effective against aphids and whiteflies. It is a selective aphicide with rapid contact and translaminar actions.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - MP - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
EZ9100000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?