EMAIL THIS PAGE TO A FRIEND
PS775 Supelco

Methomyl

analytical standard

  • CAS Number 16752-77-5

  • Empirical Formula (Hill Notation) C5H10N2O2S

  • Molecular Weight 162.21

  •  Beilstein/REAXYS Number 3081564

  •  EC Number 240-815-0

  •  MDL number MFCD00055451

Purchase

Properties

grade   analytical standard
InChI Key   UHXUZOCRWCRNSJ-QPJJXVBHSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-775
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
AK2975000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?