EMAIL THIS PAGE TO A FRIEND
PS813 Supelco

Endosulfan sulfate

analytical standard

  • CAS Number 1031-07-8

  • Empirical Formula (Hill Notation) C9H6Cl6O4S

  • Molecular Weight 422.92

  •  MDL number MFCD00151179

  •  PubChem Substance ID 24899298

Purchase

Properties

grade   analytical standard
InChI Key   AAPVQEMYVNZIOO-WINLOITPSA-N
packaging   ampule of 50 mg
mfr. no.   Chem Service, Inc. PS-81-3
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 2 - EHS - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
RB9150000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?