EMAIL THIS PAGE TO A FRIEND
PS86 Supelco

Malathion

analytical standard

  • CAS Number 121-75-5

  • Empirical Formula (Hill Notation) C10H19O6PS2

  • Molecular Weight 330.36

  •  Beilstein/REAXYS Number 1804525

  •  EC Number 204-497-7

  •  MDL number MFCD00036233

  •  PubChem Substance ID 24899302

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 500 mg
mfr. no.   Chem Service, Inc., PS-86
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC
InChI   1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3
InChI key   JXSJBGJIGXNWCI-UHFFFAOYSA-N

Description

General description

Malathion is a non-systemic pesticide belonging to the organophosphates group of pesticides. It is widely used in controlling mosquitoes, medfly, household insects, animal parasites in field crops, livestock, agriculture and fruit trees.

Application

Malathion may be used as a reference standard in the determination of malathion in pesticide formulations using fourier transform infrared (FTIR) spectrometry and attenuated total reflectance-fourier transform infrared (ATR-FTIR) spectrometry.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
WM8400000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?