EMAIL THIS PAGE TO A FRIEND
PS93 Supelco

EPN

analytical standard

Synonym: O-Ethyl O-(4-nitrophenyl) phenylphosphonothioate

  • CAS Number 2104-64-5

  • Empirical Formula (Hill Notation) C14H14NO4PS

  • Molecular Weight 323.30

  •  Beilstein/REAXYS Number 2542580

  •  EC Number 218-276-8

  •  MDL number MFCD00055309

Purchase

Properties

grade   analytical standard
InChI Key   AIGRXSNSLVJMEA-UHFFFAOYSA-N
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-93
Featured Industry   Agriculture
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGI
WGK Germany 
3
RTECS 
TB1925000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?