• USA Home
  • BP805 - Ondansetron for TLC system suitability

EMAIL THIS PAGE TO A FRIEND
BP805

Ondansetron for TLC system suitability

British Pharmacopoeia (BP) Reference Standard

Synonym: 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one hydrochloride, GR 38032F, Ondansetron hydrochloride dihydrate

  • CAS Number 103639-04-9

  • Empirical Formula (Hill Notation) C18H19N3O · HCl · 2H2O

  • Molecular Weight 365.85

  •  MDL number MFCD00374371

Purchase

Properties

grade   pharmaceutical primary standard
mfr. no.   BP
storage temp.   2-8°C
SMILES string   Cl[H].[H]O[H].[H]O[H].Cc1nccn1CC2CCc3c(C2=O)c4ccccc4n3C
InChI   1S/C18H19N3O.ClH.2H2O/c1-12-19-9-10-21(12)11-13-7-8-16-17(18(13)22)14-5-3-4-6-15(14)20(16)2;;;/h3-6,9-10,13H,7-8,11H2,1-2H3;1H;2*1H2
InChI key   VRSLTNZJOUZKLX-UHFFFAOYSA-N
Gene Information   human ... HTR3A(3359)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

5-HT3 serotonin receptor antagonist

Packaging

Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
WGK Germany 
WGK 3
RTECS 
FE6375500
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?