• USA Home
  • CRM40888 - Hydrocarbon Test Mix

EMAIL THIS PAGE TO A FRIEND
CRM40888 Supelco

Hydrocarbon Test Mix

certified reference material, 2 % (w/w) each component in isooctane

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Column & Instrument Performance Test Mixes,
grade   certified reference material
  TraceCERT®
packaging   ampule of 1 mL
concentration   2 % (w/w) each component in isooctane
application(s)   gas chromatography (GC): suitable
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Decane 2 % (w/w)    
Heptane 2 % (w/w)    
2-Methylheptane 2 % (w/w)    
4-Methylheptane 2 % (w/w)    
Octane 2 % (w/w)    
Pentane 2 % (w/w)    
Toluene 2 % (w/w)    
Safety & Documentation

Safety Information

RIDADR 
UN1993 - class 3 - PG 2 - EHS - VP <= 110kPa - Flammable liquids, n.o.s., HI: all

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?