• USA Home
  • CRM47180 - Methacrolein-2,4-DNPH Solution

EMAIL THIS PAGE TO A FRIEND
CRM47180 Supelco

Methacrolein-2,4-DNPH Solution

certified reference material, 100 μg/mL in acetonitrile, ampule of 1 mL

Synonym: Methacrolein-2,4-dinitrophenylhydrazone solution

Purchase

Properties

Related Categories Additional Standards, Air Monitoring, Air Monitoring Standards, Air Pollutants, Aldehydes / DNPH Standards, Aldehyde- and Ketone-DNPHs,
Quality Level   100
grade   certified reference material
  TraceCERT®
packaging   ampule of 1 mL
concentration   100 μg/mL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   single component solution
storage temp.   2-8°C
InChI   1S/C10H10N4O4/c1-7(2)6-11-12-9-4-3-8(13(15)16)5-10(9)14(17)18/h3-6,12H,1H2,2H3/b11-6+
InChI key   TUBWWZMXYOKKNG-IZZDOVSWSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of 13 Aldehyde/Ketone DNPH Derivatives on Ascentis® Express C18 (High Flow Rate)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 13 Aldehyde/Ketone DNPH Derivatives on Ascentis® Express C18 (High Flow Rate)
Keywords: Air Monitoring, Chromatography, High performance liquid chromatography

HPLC Analysis of 13 Aldehyde/Ketone DNPH Derivatives on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 13 Aldehyde/Ketone DNPH Derivatives on Ascentis® Express C18
Keywords: Air Monitoring, Chromatography, High performance liquid chromatography

UHPLC Analysis of Aldehyde and Ketone DNPH Derivatives on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Aldehyde and Ketone DNPH Derivatives on Ascentis® Express C18
Keywords: Air Monitoring, Chromatography, Environmental, Separation

Related Products

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?