• USA Home
  • ES5362 - GreenSep Pyridyl Amide SFC 5 Micron HPLC Column

EMAIL THIS PAGE TO A FRIEND
ES5362

GreenSep Pyridyl Amide SFC 5 Micron HPLC Column

5 μm particle size, L × I.D. 25 cm × 10 mm

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, GreenSep Pryidyl Amide 5 Micron SFC Column, GreenSep Pyridyl Amide SFC HPLC Columns, HPLC/UHPLC, SFC (Supercritical Fluid Chromatography) Columns More...
Quality Level   100
mfr. no.   ES Industries, 157291-GS-PYA
application(s)   HPLC: suitable
L × I.D.   25 cm × 10 mm
matrix active group   pyridyl amide phase
particle size   5 μm
pore size   120 Å pore size
separation technique   supercritical fluid (SFC)

Description

General description

A metal free, ultra high purity chromatographic media that is pressure stable and specifically engineered for high performance SFC separations. The surface is treated to produce optimum SFC separation interactions and loading capacity while maintaining superior peak shape perfor- mance for many chemicals. GreenSep Silica is available for analytical and preparative column formats in particle sizes from 1.8 μm to 20 μm.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?